1-Bromo-4-chloro-2-iodobenzene 98%
Supplier: Apollo Scientific
Synonyms:
2-Bromo-5-chloroiodobenzene, 1-Bromo-4-Chloro-2-Iodobenzene
OR400650-5G
OR400650-25G
APOSOR400650-5GEA
59.1
GBP
APOSOR400650-5G
APOSOR400650-25G
1-Bromo-4-chloro-2-iodobenzene 98%
1-Bromo-4-chloro-2-iodobenzene
Formula:
C₆H₃BrCl MW: 317,3495 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00672943 CAS Number: 148836-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...