3,4-Dibromoanisole

Supplier: Apollo Scientific

Synonyms: 1,2-Dibromo-4-methoxybenzene

OR400564-1G OR400564-5G
APOSOR400564-1GEA 44.6 GBP
APOSOR400564-1G APOSOR400564-5G
3,4-Dibromoanisole
3,4-Dibromoanisole
Formula: C₇H₆Br₂O
MW: 265,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070786
CAS Number: 62415-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR