3-(Benzyloxy)benzoic acid

Supplier: Apollo Scientific

OR400474-1G OR400474-5G
APOSOR400474-1GEA 22 GBP
APOSOR400474-1G APOSOR400474-5G
3-(Benzyloxy)benzoic acid
3-(Benzyloxy)benzoic acid
Formula: C₁₄H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00527901
CAS Number: 69026-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR