2-Amino-6-chlorophenol

Supplier: Apollo Scientific
Warning

OR400447-25G OR400447-5G
APOSOR400447-25GEA 254 GBP
APOSOR400447-25G APOSOR400447-5G
2-Amino-6-chlorophenol
2-Amino-6-chlorophenol
Formula: C₆H₆ClNO
MW: 143.5700 g/mol
Melting Pt: 80…81 °C
Flash Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD07801210
CAS Number: 38191-33-2
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR