3-Bromo-5-chlorobenzaldehyde 95%
Supplier: Apollo Scientific
OR400323-10G
OR400323-1G
OR400323-5G
APOSOR400323-10GEA
168
GBP
APOSOR400323-10G
APOSOR400323-1G
APOSOR400323-5G
3-Bromo-5-chlorobenzaldehyde 95%
3-Bromo-5-chlorobenzaldehyde
Formula:
C₇H₄BrClO MW: 219,4600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797226 CAS Number: 188813-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...