4-(Imidazol-1-yl)benzylamine 95+%
Supplier: Apollo Scientific
OR40029-1G
OR40029-10G
OR40029-5G
APOSOR40029-1GEA
247
GBP
APOSOR40029-1G
APOSOR40029-10G
APOSOR40029-5G
4-(Imidazol-1-yl)benzylamine 95+%
4-(Imidazol-1-yl)benzylamine
Formula:
C₁₀H₁₁N₃ MW: 173,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06213154 CAS Number: 65113-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...