2'-Bromo-4'-chloroacetophenone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(2-Bromo-4-chlorophenyl)ethanone

OR400212-5G OR400212-1G
APOSOR400212-5GEA 165 GBP
APOSOR400212-5G APOSOR400212-1G
2'-Bromo-4'-chloroacetophenone
2'-Bromo-4'-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233,4896 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683063
CAS Number: 825-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR