4-Bromo-3-nitrobenzaldehyde

Supplier: Apollo Scientific

OR400141-25G OR400141-5G
APOSOR400141-25GEA 119 GBP
APOSOR400141-25G APOSOR400141-5G
4-Bromo-3-nitrobenzaldehyde
4-Bromo-3-nitrobenzaldehyde
Formula: C₇H₄BrNO₃
MW: 230,02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00204031
CAS Number: 163596-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR