2-Methoxy-5-nitrobenzaldehyde
Supplier: Apollo Scientific
Synonyms:
5-Nitro-O-anisaldehyde
OR400049-1G
OR400049-5G
APOSOR400049-1GEA
55.1
GBP
APOSOR400049-1G
APOSOR400049-5G
2-Methoxy-5-nitrobenzaldehyde
2-Methoxy-5-nitrobenzaldehyde
Formula:
C₈H₇NO₄ MW: 181,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02093683 CAS Number: 25016-02-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...