3-(Butylcarbamoyl)phenylboronic acid 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(Butylaminocarbonyl)phenylboronic acid, 3-(n-Butylcarbamoyl)benzeneboronic acid, (3-(Butylcarbamoyl)phenyl)boronic acid

OR3980-1G OR3980-5G
APOSOR3980-1GEA 114 GBP
APOSOR3980-1G APOSOR3980-5G
3-(Butylcarbamoyl)phenylboronic acid 97%
3-(Butylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221,06 g/mol
Melting Pt: 216…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115691
CAS Number: 397843-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR