4-Cyano-2-pyridineboronic acid 98%

Supplier: Apollo Scientific

OR361048-1G OR361048-250MG
APOSOR361048-1GEA 851 GBP
APOSOR361048-1G APOSOR361048-250MG
4-Cyano-2-pyridineboronic acid 98%
4-Cyano-2-pyridineboronic acid
Formula: C₆H₅BN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD08703250
CAS Number: 1072946-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR