2-Morpholino-5-pyridineboronic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Morpholino)pyridine-5-boronic acid, 6-(Morpholino)pyridine-3-boronic acid, 6-Morpholinopyridine-3-boronic acid, 6-Morpholino-3-pyridineboronic Acid
OR360316-1G
OR360316-25G
OR360316-5G
APOSOR360316-1GEA
25
GBP
APOSOR360316-1G
APOSOR360316-25G
APOSOR360316-5G
2-Morpholino-5-pyridineboronic acid 98%
2-Morpholino-5-pyridineboronic acid
Formula:
C₉H₁₃BN₂O₃ MW: 208.0200 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD03086116 CAS Number: 904326-93-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...