2-Chloro-5-pyrimidineboronic acid

Supplier: Apollo Scientific
Warning

Synonyms: 2-Chloropyrimidine-5-boronic acid

OR360157-25G OR360157-5G
APOSOR360157-25GEA 104 GBP
APOSOR360157-25G APOSOR360157-5G
2-Chloro-5-pyrimidineboronic acid
2-Chloro-5-pyrimidineboronic acid
Formula: C₄H₄BClN₂O₂
MW: 158,35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063113
CAS Number: 1003845-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR