(Imidazo[2,1-b]benzothiazol-2-yl)methanol 95+%
Supplier: Apollo Scientific
|
Warning
|
OR3587-1G
OR3587-25G
OR3587-5G
APOSOR3587-1GEA
189
GBP
APOSOR3587-1G
APOSOR3587-25G
APOSOR3587-5G
(Imidazo[2,1-b]benzothiazol-2-yl)methanol 95+%
(Imidazo[2,1-b]benzothiazol-2-yl)methanol
Formula:
C₁₀H₈N₂OS MW: 204,25 g/mol Melting Pt: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00203010 CAS Number: 114095-02-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...