3-Bromo-2,5-dichloropyridine

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-2,5-dichloro-pyridine

OR3581-5G OR3581-1G OR3581-25G
APOSOR3581-5GEA 31.5 GBP
APOSOR3581-5G APOSOR3581-1G APOSOR3581-25G
3-Bromo-2,5-dichloropyridine
3-Bromo-2,5-dichloropyridine
Formula: C₅H₂BrCl₂N
MW: 226,89 g/mol
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD06659484
CAS Number: 138006-41-4
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR