6-Bromo[1,2,4]triazolo[1,5-a]pyridine 99%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine
OR3552-1G
OR3552-5G
OR3552-25G
APOSOR3552-1GEA
62.9
GBP
APOSOR3552-1G
APOSOR3552-5G
APOSOR3552-25G
6-Bromo[1,2,4]triazolo[1,5-a]pyridine 99%
6-Bromo-[1,2,4]triazolo[1,5-a]pyridine
Formula:
C₆H₄BrN₃ MW: 198,02 g/mol Melting Pt: 115…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08689534 CAS Number: 356560-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...