Methyl-5-bromo-2-chloropyridine-4-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 5-bromo-2-chloroisonicotinate

OR3519-1G OR3519-5G
APOSOR3519-1GEA 80 GBP
APOSOR3519-1G APOSOR3519-5G
Methyl-5-bromo-2-chloropyridine-4-carboxylate
Methyl-5-bromo-2-chloropyridine-4-carboxylate
Formula: C₇H₅BrClNO₂
MW: 250,49 g/mol
Melting Pt: 31…34 °C
Storage Temperature: Ambient
MDL Number: MFCD07375113
CAS Number: 886365-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR