3,4,7,8-Tetramethyl-1,10-phenanthroline 95%
Supplier: Apollo Scientific
OR350386-25G
OR350386-5G
APOSOR350386-25GEA
197
GBP
APOSOR350386-25G
APOSOR350386-5G
3,4,7,8-Tetramethyl-1,10-phenanthroline 95%
3,4,7,8-Tetramethyl-1,10-phenanthroline
Formula:
C₁₆H₁₆N₂ MW: 236,32 g/mol Melting Pt: 277…280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004974 CAS Number: 1660-93-1 EINECS: 216-762-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...