Thiazole 98%
Supplier: Apollo Scientific
|
Danger
|
OR350371-25G
OR350371-5G
APOSOR350371-25GEA
131
GBP
APOSOR350371-25G
APOSOR350371-5G
Thiazole 98%
Thiazole
Formula:
C₃H₃NS MW: 85.13 g/mol Boiling Pt: 117…118 °C/760mmHg Melting Pt: –33 °C Density: 1.198 g/cm³ (25 °C) Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005315 CAS Number: 288-47-1 UN: 1993 ADR: 3 III Merck Index: 12,09442 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...