Phenyl vinyl sulphone 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Vinyl phenyl sulphone, (Vinylsulphonyl)benzene
OR350320-25G
OR350320-1G
OR350320-5G
APOSOR350320-25GEA
59.1
GBP
APOSOR350320-25G
APOSOR350320-1G
APOSOR350320-5G
Phenyl vinyl sulphone 97%
Phenyl vinyl sulphone
Formula:
C₈H₈O₂S MW: 168,22 g/mol Melting Pt: 68…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007554 CAS Number: 5535-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...