2-Isopropyl-4-methylthiazole
Supplier: Apollo Scientific
|
Warning
|
OR350201-100G
OR350201-25G
OR350201-500G
APOSOR350201-100GEA
38
GBP
APOSOR350201-100G
APOSOR350201-25G
APOSOR350201-500G
2-Isopropyl-4-methylthiazole
2-Isopropyl-4-methylthiazole
Formula:
C₇H₁₁NS Boiling Pt: 92 °C Density: 1,001 g/cm³ (25 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036561 CAS Number: 15679-13-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...