Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan

Supplier: Apollo Scientific

Synonyms: Fmoc-L-Trp(Boc)-OH, N1-Boc-Nɑ-Fmoc-L-tryptophan, 1-Boc-N-Fmoc-L-tryptophan, Fmoc-Trp(Boc)-OH

OR350178-25G OR350178-5G OR350178-10G
APOSOR350178-25GEA 91.8 GBP
APOSOR350178-25G APOSOR350178-5G APOSOR350178-10G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan
Formula: C₃₁H₃₀N₂O₆
MW: 526,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153366
CAS Number: 143824-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR