(3-Isopropylisoxazol-5-yl)methanol 95+%
Supplier: Apollo Scientific
OR346706-1G
OR346706-5G
APOSOR346706-1GEA
287
GBP
APOSOR346706-1G
APOSOR346706-5G
(3-Isopropylisoxazol-5-yl)methanol 95+%
(3-Isopropylisoxazol-5-yl)methanol
Formula:
C₇H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD10001572 CAS Number: 14633-17-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...