3-tert-Butyl-1H-pyrazole-5-carbaldehyde 95+%

Supplier: Apollo Scientific

OR346539-1G OR346539-5G
APOSOR346539-1GEA 165 GBP
APOSOR346539-1G APOSOR346539-5G
3-tert-Butyl-1H-pyrazole-5-carbaldehyde 95+%
3-tert-Butyl-1H-pyrazole-5-carbaldehyde
Formula: C₈H₁₂N₂O
MW: 152,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12028432
CAS Number: 865138-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR