N-(3-Methoxyphenyl)acetamide 95+%
Supplier: Apollo Scientific
OR346506-1G
OR346506-5G
APOSOR346506-1GEA
85.2
GBP
APOSOR346506-1G
APOSOR346506-5G
N-(3-Methoxyphenyl)acetamide 95+%
N-(3-Methoxyphenyl)acetamide
Formula:
C₉H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00123297 CAS Number: 588-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...