Formanilide 95+%

Supplier: Apollo Scientific

Synonyms: N-Phenyl-formamide

OR346451-10G OR346451-1G OR346451-5G
APOSOR346451-10GEA 477 GBP
APOSOR346451-10G APOSOR346451-1G APOSOR346451-5G
Formanilide 95+%
Formanilide
Formula: C₇H₇NO
MW: 121,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003276
CAS Number: 103-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR