Phenoxyacetone 95+%

Supplier: Apollo Scientific

Synonyms: 1-Phenoxypropan-2-one

OR346450-10G OR346450-1G OR346450-5G
APOSOR346450-10GEA 477 GBP
APOSOR346450-10G APOSOR346450-1G APOSOR346450-5G
Phenoxyacetone 95+%
Phenoxyacetone
Formula: (C₆H₅)OCH₂C(O)CH₃
MW: 150,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008767
CAS Number: 621-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR