3-Oxazol-5-ylbenzenesulphonyl chloride

Supplier: Apollo Scientific
Danger

OR346098-1G OR346098-500MG
APOSOR346098-1GEA 0 GBP
APOSOR346098-1G APOSOR346098-500MG
3-Oxazol-5-ylbenzenesulphonyl chloride
3-Oxazol-5-ylbenzenesulphonyl chloride
Formula: C₉H₆ClNO₃S
MW: 243,67 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD03407346
CAS Number: 499770-75-1
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR