tert-Butyl-2-formylbenzoate

Supplier: Apollo Scientific

OR346067-1G OR346067-250MG
APOSOR346067-1GEA 231 GBP
APOSOR346067-1G APOSOR346067-250MG
tert-Butyl-2-formylbenzoate
tert-Butyl-2-formylbenzoate
Formula: C₁₂H₁₄O₃
MW: 206,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02180884
CAS Number: 907948-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR