tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: 1-tert-Butyloxycarbonyl-3-bromo-4-oxopiperidine, tert-Butyl 3-bromo-4-oxopiperidine-1-carboxylate

OR346051-1G OR346051-5G
APOSOR346051-1GEA 46 GBP
APOSOR346051-1G APOSOR346051-5G
tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate
tert-Butyl-3-bromo-4-oxopiperidine-1-carboxylate
Formula: C₁₀H₁₆BrNO₃
MW: 278,1429 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD06738530
CAS Number: 188869-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR