Pyrazine-2-N-cyanoamidine
Supplier: Apollo Scientific
OR346029-1G
OR346029-5G
APOSOR346029-1GEA
0
GBP
APOSOR346029-1G
APOSOR346029-5G
Pyrazine-2-N-cyanoamidine
Pyrazine-2-N-cyanoamidine
Formula:
C₆H₅N₅ Storage Temperature: Ambient |
MDL Number:
MFCD09864979 CAS Number: 1053656-81-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...