4-Chloro-2-nitropyridine 97%
Supplier: Apollo Scientific
|
Warning
|
OR345390-1G
OR345390-5G
OR345390-10G
APOSOR345390-1GEA
52.5
GBP
APOSOR345390-1G
APOSOR345390-5G
APOSOR345390-10G
4-Chloro-2-nitropyridine 97%
4-Chloro-2-nitropyridine
Formula:
C₅H₃ClN₂O₂ Melting Pt: 46…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114167 CAS Number: 65370-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...