5-Bromo-4-chloro-2-(methylthio)pyrimidine

Supplier: Apollo Scientific
Danger

Synonyms: 5-bromo-4-chloro-2-(methylsulphanyl)pyrimidine

OR345386-25G OR345386-5G
APOSOR345386-25GEA 130 GBP
APOSOR345386-25G APOSOR345386-5G
5-Bromo-4-chloro-2-(methylthio)pyrimidine
5-Bromo-4-chloro-2-(methylthio)pyrimidine
Formula: C₅H₄BrClN₂S
MW: 239,5200 g/mol
Melting Pt: 45…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00023232
CAS Number: 63810-78-6
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR