6-Bromo-2-nitropyridin-3-ol 97%
Supplier: Apollo Scientific
Synonyms:
6-Bromo-3-hydroxy-2-nitropyridine
OR345348-1G
OR345348-5G
APOSOR345348-1GEA
61.7
GBP
APOSOR345348-1G
APOSOR345348-5G
6-Bromo-2-nitropyridin-3-ol 97%
6-Bromo-2-nitropyridin-3-ol
Formula:
C₅H₃BrN₂O₃ MW: 218,9900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261204 CAS Number: 443956-08-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...