2-(Benzyloxy)benzoic acid
Supplier: Apollo Scientific
OR345143-25G
OR345143-5G
APOSOR345143-25GEA
94.2
GBP
APOSOR345143-25G
APOSOR345143-5G
2-(Benzyloxy)benzoic acid
2-(Benzyloxy)benzoic acid
Formula:
C₁₄H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00051940 CAS Number: 14389-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...