Pyrazine-2-carboximidamide hydrochloride 97%

Supplier: Apollo Scientific
Warning

OR3402-10G OR3402-5G
APOSOR3402-10GEA 57.7 GBP
APOSOR3402-10G APOSOR3402-5G
Pyrazine-2-carboximidamide hydrochloride 97%
Pyrazine-2-carboximidamide hydrochloride
Formula: C5H7ClN4
MW: 158,5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663360
CAS Number: 138588-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR