2-Chloro-N-phenylacetamide 97%

Supplier: Apollo Scientific
Warning

Synonyms: N-(Chloroacetyl)aniline

OR3397-5G OR3397-1G
APOSOR3397-5GEA 59.1 GBP
APOSOR3397-5G APOSOR3397-1G
2-Chloro-N-phenylacetamide 97%
2-Chloro-N-phenylacetamide
Formula: C₈H₈ClNO
MW: 169,61 g/mol
Melting Pt: 136…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00018887
CAS Number: 587-65-5
EINECS: 209-604-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR