Pyrimidine 98%
Supplier: Apollo Scientific
|
Warning
|
OR33655-100G
OR33655-25G
APOSOR33655-100GEA
683
GBP
APOSOR33655-100G
APOSOR33655-25G
Pyrimidine 98%
Pyrimidine
Formula:
C₄H₄N₂ MW: 80,09 g/mol Boiling Pt: 123…124 °C Melting Pt: 19…22 °C Density: 1,016 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006059 CAS Number: 289-95-2 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...