Ethyl-5-methylpicolinate 97%
Supplier: Apollo Scientific
Synonyms:
Ethyl 5-methylpicolinate, Ethyl 5-methylpyridine-2-carboxylate
OR33564-1G
OR33564-5G
OR33564-25G
APOSOR33564-1GEA
163
GBP
APOSOR33564-1G
APOSOR33564-5G
APOSOR33564-25G
Ethyl-5-methylpicolinate 97%
Ethyl 5-methylpicolinate
Formula:
C₉H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD10566273 CAS Number: 55876-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...