2-Chloro-6-nitrobenzoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
OR3351-5G
OR3351-25G
APOSOR3351-5GEA
13.2
GBP
APOSOR3351-5G
APOSOR3351-25G
2-Chloro-6-nitrobenzoic acid 98%
2-Chloro-6-nitrobenzoic acid
Formula:
C₇H₄ClNO₄ MW: 201,57 g/mol Melting Pt: 160…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00100425 CAS Number: 5344-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...