1-Ethynyl-2-methoxy-4-nitrobenzene 95%
Supplier: Apollo Scientific
OR33492-1G
OR33492-5G
APOSOR33492-1GEA
400
GBP
APOSOR33492-1G
APOSOR33492-5G
1-Ethynyl-2-methoxy-4-nitrobenzene 95%
1-Ethynyl-2-methoxy-4-nitrobenzene
Formula:
C₉H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD11840985 CAS Number: 1135283-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...