N'-(1H-Indazol-5-yl)-N,N-dimethyliminoformamide 95%

Supplier: Apollo Scientific

Synonyms: (E)-N'-(1H-Indazol-5-yl)-N,N-dimethylmethanimidamide

OR33465-1G OR33465-5G
APOSOR33465-1GEA 211 GBP
APOSOR33465-1G APOSOR33465-5G
N'-(1H-Indazol-5-yl)-N,N-dimethyliminoformamide 95%
N'-(1H-Indazol-5-yl)-N,N-dimethyliminoformamide
Formula: C₁₀H₁₂N₄
MW: 188,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09152720
CAS Number: 952183-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR