2-(4-Ethylpiperazino)acetonitrile 95%
Supplier: Apollo Scientific
OR33463-1G
OR33463-5G
APOSOR33463-1GEA
243
GBP
APOSOR33463-1G
APOSOR33463-5G
2-(4-Ethylpiperazino)acetonitrile 95%
2-(4-Ethylpiperazino)acetonitrile
Formula:
C₈H₁₅N₃ MW: 153.23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09152768 CAS Number: 90206-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...