N'-(1H-Indazol-6-yl)-N,N-dimethylethanimidamide 95%

Supplier: Apollo Scientific

OR33461-1G OR33461-5G
APOSOR33461-1GEA 211 GBP
APOSOR33461-1G APOSOR33461-5G
N'-(1H-Indazol-6-yl)-N,N-dimethylethanimidamide 95%
N'-(1H-Indazol-6-yl)-N,N-dimethylethanimidamide
Formula: C₁₁H₁₄N₄
MW: 202,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09152717
CAS Number: 952183-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR