Ethyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate 95%

Supplier: Apollo Scientific

OR33414-1G OR33414-5G
APOSOR33414-1GEA 151 GBP
APOSOR33414-1G APOSOR33414-5G
Ethyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate 95%
Ethyl-1-(6-chloro-3-pyridazinyl)-4-piperidinecarboxylate
Formula: C₁₂H₁₆ClN₃O₂
MW: 269,73 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425828
CAS Number: 252263-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR