{1-[2-(Aminomethyl)phenyl]-4-piperidinyl}methanol 95%
Supplier: Apollo Scientific
OR33411-1G
OR33411-5G
APOSOR33411-1GEA
269
GBP
APOSOR33411-1G
APOSOR33411-5G
{1-[2-(Aminomethyl)phenyl]-4-piperidinyl}methanol 95%
{1-[2-(Aminomethyl)phenyl]-4-piperidinyl}methanol
Formula:
C₁₃H₂₀N₂O MW: 220,32 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425826 CAS Number: 220060-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...