2-Oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile 95%

Supplier: Apollo Scientific

OR33351-1G OR33351-5G
APOSOR33351-1GEA 211 GBP
APOSOR33351-1G APOSOR33351-5G
2-Oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile 95%
2-Oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile
Formula: C₁₂H₈N₂O
MW: 196,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00202791
CAS Number: 43083-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR