4-(4-Methoxyphenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 95%

Supplier: Apollo Scientific

OR33332-1G OR33332-25G OR33332-5G
APOSOR33332-1GEA 159 GBP
APOSOR33332-1G APOSOR33332-25G APOSOR33332-5G
4-(4-Methoxyphenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile 95%
4-(4-Methoxyphenyl)-2-oxo-2,5-dihydro-3-furancarbonitrile
Formula: C₁₂H₉NO₃
MW: 215,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00139243
CAS Number: 50691-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR