4'-Chloro-2-cyanoacetanilide 95%

Supplier: Apollo Scientific

Synonyms: N-(4-Chlorophenyl)-2-cyanoacetamide

OR33321-25G OR33321-5G
APOSOR33321-25GEA 545 GBP
APOSOR33321-25G APOSOR33321-5G
4'-Chloro-2-cyanoacetanilide 95%
4'-Chloro-2-cyanoacetanilide
Formula: C₉H₇ClN₂O
MW: 194,6177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036015
CAS Number: 17722-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR