2-Isopropylpiperazine 95%

Supplier: Apollo Scientific

OR33314-1G OR33314-5G
APOSOR33314-1GEA 269 GBP
APOSOR33314-1G APOSOR33314-5G
2-Isopropylpiperazine 95%
2-Isopropylpiperazine
Formula: C₇H₁₆N₂
MW: 128,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07373042
CAS Number: 84468-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR